Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 4G4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF502930
ORIGCF502930EA 833 EUR
ORIGCF502930
Anti-RPS6KB1 Mouse Monoclonal Antibody [clone: 4G4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RPS6KB1
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool RPS6KB1
Environmentally Preferable
Type antilichaam Primair
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone 4G4
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Epitoop Full length
Immunogeen Full length human recombinant protein of human RPS6KB1 (NP_003152) produced in HEK293T cell.
Formulier Lyophilized
Moleculair gewicht 59 kDa
Purification purified by affinity chromatography
Kruisadsorptie No
Opslagbuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Opslagtemperatuur –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR