Anti-TIMP2 Mouse Monoclonal Antibody [clone: 3H8]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF504042
ORIGCF504042EA 833 EUR
ORIGCF504042
Anti-TIMP2 Mouse Monoclonal Antibody [clone: 3H8]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TIMP2
Clonality: Monoclonal
Clone: 3H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR