Anti-S100P Mouse Monoclonal Antibody [clone: 1A4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF504478
ORIGCF504478EA 833 EUR
ORIGCF504478
Anti-S100P Mouse Monoclonal Antibody [clone: 1A4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: S100P
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool S100P
Environmentally Preferable
Type antilichaam Primair
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone 1A4
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoFluorescentie Yes
Epitoop Full length
Immunogeen Full length human recombinant protein of human S100P(NP_005971) produced in HEK293T cell.
Formulier Lyophilized
Moleculair gewicht 10.2 kDa
Purification purified by affinity chromatography
Kruisadsorptie No
Opslagbuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Opslagtemperatuur –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR