Anti-CHN1 Mouse Monoclonal Antibody [clone: 1B7]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF505073
ORIGCF505073EA 833 EUR
ORIGCF505073
Anti-CHN1 Mouse Monoclonal Antibody [clone: 1B7]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: CHN1
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR