Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G3]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CF505890
ORIGCF505890EA 833 EUR
ORIGCF505890
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 2G3]
Antilichamen
Recommended Dilutions: Western Blot: 1:200-2000; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 2G3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR