Slc7a11 (NM_011990) mouse tagged ORF clone

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MG225346
ORIGMG225346EA 1260 EUR
ORIGMG225346
Slc7a11 (NM_011990) mouse tagged ORF clone
Vectoren, Plasmiden en Bibliotheken

Slc7a11 (GFP-tagged) - Mouse solute carrier family 7 (cationic amino acid transporter y+ system) member 11 (Slc7a11), (10 µg).


  • Type: Mouse tagged ORF clone
  • Tag: TurboGFP
  • Symbol: Slc7a11
  • Synonyms: 9930009M05Rik; AI451155; sut; xCT
  • Vector: pCMV6-AC-GFP
  • E. coli Selection: Ampicillin (100 µg/ml)
  • Mammalian cell selection: Neomycin
  • Restriction sites: SgfI-MluI

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR