SLK Human siRNA Oligo Duplex (Locus ID 9748)
Leverancier: OriGene
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
SR306513
ORIGSR306513EA 593 EUR
ORIGSR306513
SLK Human siRNA Oligo Duplex (Locus ID 9748)
Reagentia voor nucleïnezuren RNAi
This SLK Human siRNA Oligo Duplex mediates apoptosis and actin stress fiber dissolution.
  • HPLC purified
  • Tested by ESI-MS
Can be used for approximately 330 transfections/2 nmol in 24-well plate under optimised conditions (final conc. 10 nM).
Leveringsinformatie: Kit contains SLK (Human) - 3 unique 27mer siRNA duplexes - 2 nmol each (Locus ID 9748), Trilencer-27 universal scrambled negative control siRNA duplex - 2 nmol, RNAse free siRNA duplex resuspension buffer - 2 ml.