Anti-FBXO32 Goat antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA305697
ORIGTA305697EA 785 EUR
ORIGTA305697
Anti-FBXO32 Goat antibody
Antilichamen
Recommended Dilutions: Western Blot: 01-03?g/ml

Type: Primary
Antigen: FBXO32 (F-box protein 32)
Clonality:
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR