Anti-DDX3X Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA306299
ORIGTA306299EA 792 EUR
ORIGTA306299
Anti-DDX3X Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: DDX3X (DEAD (Asp-Glu-Ala-Asp) box helicase 3, X-linked)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR