Anti-KLRG1 Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA308319
ORIGTA308319EA 868 EUR
ORIGTA308319
Anti-KLRG1 Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot:1:500-1:3000

Type: Primary
Antigen: KLRG1 (killer cell lectin-like receptor subfamily G, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR