Anti-PRKACA Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA314380
ORIGTA314380EA 528 EUR
ORIGTA314380
Anti-PRKACA Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:20000

Type: Primary
Antigen: PRKACA (protein kinase, cAMP-dependent, catalytic, alpha)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool PRKACA
Type antilichaam Primair,Protein Kinase, cAMP-dependent, Catalytic Alpha
Environmentally Preferable
Type antilichaam Primair,Protein Kinase, cAMP-dependent, Catalytic Alpha
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Formulier Liquid
Purification purified by affinity chromatography
Kruisadsorptie none
Opslagbuffer Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR