Anti-VENTX Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA318898
ORIGTA318898EA 565 EUR
ORIGTA318898
Anti-VENTX Rabbit Polyclonal Antibody
Antilichamen
Recommended Dilutions: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: VENTX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR