Anti-DLL1 Armenian Hamster Polyclonal Antibody [clone: HMD1-5]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA320321
ORIGTA320321EA 352 EUR
ORIGTA320321
Anti-DLL1 Armenian Hamster Polyclonal Antibody [clone: HMD1-5]
Antilichamen
Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone: HMD1-5
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: Mouse
Order Now

Specificaties

antigeen symbool DLL1
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Clone HMD1-5
Reactivity Mouse
Host Armenian hamster
Isotype
Formulier Liquid
Purification purified by affinity chromatography
Kruisadsorptie No
Opslagbuffer Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
Opslagtemperatuur –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR