Anti-CAMK4 Rabbit Polyclonal Antibody

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA325291
ORIGTA325291EA 528 EUR
ORIGTA325291
Anti-CAMK4 Rabbit Polyclonal Antibody
Antilichamen
  • Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200; Immunofluorescence: 1:100-1:500

Type: Primary
Antigen: CAMK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Specificaties

antigeen symbool CAMK4
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Immunogeen The antiserum was produced against A synthesized peptide derived from human CaMK4
Formulier Liquid
Moleculair gewicht 60 kDa
Purification purified by affinity chromatography
Kruisadsorptie No
Opslagbuffer Rabbit IgG in phosphate buffered saline , pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Opslagtemperatuur –20 °C
Shipping temperature –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR