Anti-PDIA3 Mouse Monoclonal Antibody [clone: Map.ERP57]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA326353
ORIGTA326353EA 833 EUR
ORIGTA326353
Anti-PDIA3 Mouse Monoclonal Antibody [clone: Map.ERP57]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: Map.ERP57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, canine, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR