Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI1C9]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA501425
ORIGTA501425EA 621 EUR
ORIGTA501425
Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI1C9]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: EFNA2 (ephrin-A2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR