Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI7C2]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA501533
ORIGTA501533EA 621 EUR
ORIGTA501533
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI7C2]
Antilichamen
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BTN1A1 (butyrophilin, subfamily 1, member A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR