Anti-KLHL2 Mouse Monoclonal Antibody [clone: OTI3H9]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA501550
ORIGTA501550EA 621 EUR
ORIGTA501550
Anti-KLHL2 Mouse Monoclonal Antibody [clone: OTI3H9]
Antilichamen
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: KLHL2 (kelch-like family member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR