Anti-OSBPL11 Mouse Monoclonal Antibody [clone: OTI5A10]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA501940
ORIGTA501940EA 621 EUR
ORIGTA501940
Anti-OSBPL11 Mouse Monoclonal Antibody [clone: OTI5A10]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: OSBPL11 (oxysterol binding protein-like 11)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool OSBPL11
Environmentally Preferable
Type antilichaam Primair,oxysterol binding Protein-like 11
Type antilichaam Primair,oxysterol binding Protein-like 11
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone OTI5A10
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
Epitoop Full length
Formulier Liquid
Moleculair gewicht 83.5 kDa
Purification purified by affinity chromatography
Kruisadsorptie none
Opslagbuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR