Anti-SERPINB4 Mouse Monoclonal Antibody [clone: OTI4C10]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA502436
ORIGTA502436EA 621 EUR
ORIGTA502436
Anti-SERPINB4 Mouse Monoclonal Antibody [clone: OTI4C10]
Antilichamen
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: SERPINB4 (serpin peptidase inhibitor, clade B (ovalbumin), member 4)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR