Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2C3]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA502761
ORIGTA502761EA 621 EUR
ORIGTA502761
Anti-NDUFA7 Mouse Monoclonal Antibody [clone: OTI2C3]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NDUFA7 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Specificaties

antigeen symbool NDUFA7
Type antilichaam Primair,NADH dehydrogenase (ubiquinone) 1, alpha subcomplex 7, 14.5kDa
Environmentally Preferable
Type antilichaam Primair,NADH dehydrogenase (ubiquinone) 1, alpha subcomplex 7, 14.5kDa
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone OTI2C3
Reactivity Human
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemie Yes
ImmunoFluorescentie Yes
Flowcytometrie Yes
Epitoop Full length
Formulier Liquid
Moleculair gewicht 12.4 kDa
Purification purified by affinity chromatography
Kruisadsorptie none
Opslagbuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR