Anti-NLN Mouse Monoclonal Antibody [clone: OTI2H1]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504180
ORIGTA504180EA 621 EUR
ORIGTA504180
Anti-NLN Mouse Monoclonal Antibody [clone: OTI2H1]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: NLN (neurolysin (metallopeptidase M3 family))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR