Anti-TFR2 Mouse Monoclonal Antibody [clone: OTI1B1]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504592
ORIGTA504592EA 621 EUR
ORIGTA504592
Anti-TFR2 Mouse Monoclonal Antibody [clone: OTI1B1]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TFR2 (transferrin receptor 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR