Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1E4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504609
ORIGTA504609EA 621 EUR
ORIGTA504609
Anti-MIEN1 Mouse Monoclonal Antibody [clone: OTI1E4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MIEN1 (migration and invasion enhancer 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR