Anti-PGM3 Mouse Monoclonal Antibody [clone: OTI1B8]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504860
ORIGTA504860EA 621 EUR
ORIGTA504860
Anti-PGM3 Mouse Monoclonal Antibody [clone: OTI1B8]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: PGM3 (phosphoglucomutase 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR