Anti-SENP2 Mouse Monoclonal Antibody [clone: OTI3H10]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA504864
ORIGTA504864EA 621 EUR
ORIGTA504864
Anti-SENP2 Mouse Monoclonal Antibody [clone: OTI3H10]
Antilichamen
Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: SENP2 (SUMO1/sentrin/SMT3 specific peptidase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR