Anti-UBE2G2 Mouse Monoclonal Antibody [clone: OTI4D4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA505238
ORIGTA505238EA 621 EUR
ORIGTA505238
Anti-UBE2G2 Mouse Monoclonal Antibody [clone: OTI4D4]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: UBE2G2 (ubiquitin-conjugating enzyme E2G 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR