Anti-CMPK1 Mouse Monoclonal Antibody [clone: OTI1D7]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA505410
ORIGTA505410EA 621 EUR
ORIGTA505410
Anti-CMPK1 Mouse Monoclonal Antibody [clone: OTI1D7]
Antilichamen
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CMPK1 (cytidine monophosphate (UMP-CMP) kinase 1, cytosolic)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Specificaties

antigeen symbool CMPK1
Type antilichaam Primair,Cytidine monophosphate (UMP-CMP) kinase 1
Environmentally Preferable
Type antilichaam Primair,Cytidine monophosphate (UMP-CMP) kinase 1
Clonality Monoklonaal
Conjugatie Onconjugeerd
Clone OTI1D7
Reactivity Human
Host Mouse
Isotype IgG2a
Western blot Yes
Epitoop Full length
Formulier Liquid
Moleculair gewicht 25.7 kDa
Purification purified by affinity chromatography
Kruisadsorptie none
Opslagbuffer PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR