Anti-ICAM3 Mouse Monoclonal Antibody [clone: 3A7]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA507175
ORIGTA507175EA 621 EUR
ORIGTA507175
Anti-ICAM3 Mouse Monoclonal Antibody [clone: 3A7]
Antilichamen
Recommended Dilutions: Western Blot: 1:400-1:2000

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR