Anti-IL17A Mouse Monoclonal Antibody [clone: OTI4C17]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA600015
ORIGTA600015EA 636 EUR
ORIGTA600015
Anti-IL17A Mouse Monoclonal Antibody [clone: OTI4C17]
Antilichamen
Recommended Dilutions: 1:100-1:1000

Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR