Anti-DEF6 Mouse Monoclonal Antibody [clone: OTI3C4]

Leverancier: OriGene
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

TA700323
ORIGTA700323EA 686 EUR
ORIGTA700323
Anti-DEF6 Mouse Monoclonal Antibody [clone: OTI3C4]
Antilichamen
  • Recommended Dilutions: 1:250-1:1000

Type: Primary
Antigen: DEF6 (differentially expressed in FDCP 6 homolog (mouse))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR