ISA-oplossingen

Leverancier: Thermo Orion
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

932011 930710 941709 950210 977010 977011 951011 940999 956410 841111 930711 931911 940011 940909 940911 941609 951211 951210
662-0818EA 224 EUR
662-0818 662-0812 662-0838 662-0851 662-0881 662-0882 ORIO951011 ORIO940999 ORIO956410 662-0745 662-0813 662-0816 662-0824 662-0828 662-0829 662-0833 662-0857 ORIO951210
ISA-oplossingen
Elektrolyten Vuloplossingen voor elektrodes

Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.


  • Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
  • Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
  • Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
  • Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences


Concentration : 10 ppm


For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR