Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices
Leverancier: Oxford Nanopore Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
ADP-FLG001 FLO-FLG114.6 FLG.SP
ADP-FLG001EA 940 EUR
ADP-FLG001 FLO-FLG114.6 OXNTFLG.SP
Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices
Doorstroomcellen
Flongle is an adapter for MinION and GridION devices that enables direct, real-time DNA sequencing on smaller, single-use Flongle Flow Cells.
  • The most rapid, accessible, and cost-efficient nanopore sequencing system for smaller or more frequently performed assays and experiments
  • Each Flongle Flow can deliver 1–2 Gb of real-time sequencing data
The Flongle Adapter allows MinION and GridION devices to run smaller, single-use Flongle Flow Cells, typically generating 1–2 Gb of data per flow cell. Users can simply insert a Flongle Flow Cell into theMore Product Information
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.