Ureum 40% aqueous solution

Leverancier: THERMO FISHER DIAGNOSTICS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide

SR0020K
OXOISR0020KEA 61.9 EUR
OXOISR0020K
Ureum 40% aqueous solution
Ureum
Formule: NH₂CONH₂
MW: 60.06 g/mol
Kookpunt: 197
Smeltpunt: 132.7
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR