Magnetic plates

Leverancier: OZ BIOSCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MF10000 MF10096 MF14000
OZBIMF10000EA 477.75 EUR
OZBIMF10000 OZBIMF10096 OZBIMF14000
Magnetic plates
Magneten
Optimised magnetic plates with tailored properties especially designed for Magnetofection™.

  • Can be easily cleaned and decontaminated with 70% ethanol
  • Can be used at room temperature, 37 °C, +4 °C, etc.
  • Compatible with culture plates from most common suppliers
  • Solid, completely reusable

Suitable for all Magnetofection™ reagents, all cell culture dishes and flasks and can be used within incubators and with robots.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR