Zilvernitraat 99.0-100.5% Ph. Eur., Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Lunar caustic, Silver mononitrate, Nitric acid silver salt

P100-31EA 0 EUR
P100-31 P100-33 P100-36 P100-42 MACRP100-38
Zilvernitraat 99.0-100.5% Ph. Eur., Macron Fine Chemicals™
Zilvernitraat
Formule: AgNO₃
MDL Nummer: MFCD00003414
CAS nummer: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Ph. Eur. - Assay 99.0 - 100.5%
Ph. Eur. - Identification A Passes Test
Ph. Eur. - Identification B Passes Test
Ph. Eur. - Appearance of Solution Passes Test
Ph. Eur. - Acidity or Alkalinity Passes Test
Ph. Eur. - Foreign Salts ≤0.3%
Ph. Eur. - Aluminium, Lead, Copper and Bismuth Passes Test
Clarity and Colour of solution Clear, Colourless
Identity A Positive Reaction
Identity B Positive Reaction
Copper Does not Appear Blue Colour
Assay 99.8 - 100.5%
For Manufacturing, Processing or ph. Eur. Repackaging
Bulk Pharmaceutical Chemical
No Class 1,2,3 or Other Solvents Are Used Or Produced In the Manufacturing or Purification of the Product
Meets Ph. Eur. Requirements


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR