Calcium carbonate Ph. Eur., Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Marble

P104-33EA 45.6 EUR
P104-33 P104-38
Calcium carbonate Ph. Eur., Macron Fine Chemicals™
Calcium carbonate
Formule: CaCO₃
MW: 100.09 g/mol
Smeltpunt: 825
MDL Nummer: MFCD00010906
CAS nummer: 471-34-1
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR