Avantor® Partisil ODS, HPLC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P10ODS-200AEA 1030 EUR
P10ODS-200A P10ODS-250AF P10ODS-50A P10ODS-10C5-SK P10ODS-300AF
Avantor® Partisil ODS, HPLC Columns
Kolommen voor chromatografie
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity for preparative work.
  • Irregular porous silica
  • Convenient and easy to use: Hand tightened fittings
  • Increased column lifetime
Low carbon load C18 phase, with intentionally unbonded surface silanols adding to the selectivity.
Opgelet: *Use with freestanding holder HI-161 and column coupler HI-081, please see "HPLC Guard Column Holders and Accessories"