Percoll®, Siam
Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P1644-1L P1644-25ML P1644-500ML P1644-100ML
P1644-1LEA 1060 EUR
P1644-1L P1644-25ML P1644-500ML P1644-100ML
Percoll®, Siam
Media voor celscheiding
Percoll® is a classic medium for density gradient centrifugation of cells, viruses, and subcellular particles. It consists of colloidal silica particles of 15 – 30 nm diameter (23% w/w in water), coated with polyvinylpyrrolidone (PVP).
  • Non toxic
  • Ideal for use with biological materials