Phosphate Buffer, pH 6.6 at 25 °C

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P8165-12VL P8165-1VL
P8165-12VLEA 283 EUR
P8165-12VL P8165-1VL
Phosphate Buffer, pH 6.6 at 25 °C
Buffers Buffers voor elektroforese Wasbuffers

Phosphate buffer's buffering capacity is pH sensitive and is preferred to be used at physiological pH (7,2 to 7,4).


  • Each vial will prepare 3,8 L of buffer, 34 mM
  • Used in hematology and histology


For use with Wright Stain for Geometric Data HEMASTAIN®. Phosphate buffer serves as an important fixative for light and electron microscopy. It is the most commonly used component of irrigation solution during surgeries, to preserve living tissues. It is also used in immunochemical staining procedures.

Order Now

Specificaties

Environmentally Preferable
Uiterlijk White powder
Opslagtemperatuur RT
M-Clarity quality level MQ200

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR