Test tubes, for autosamplers
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N0777156 N0777159 N0777167 N0777168 N9301205 B0193233 B0193234 B0508901 N0777701 N0777702 N0777703 N0777704 N0777705 N0777706 N0777692 N0777693 N0777695 N0777696 N0777697 N0777698 N0777699 N0777700 N0777940 N0777694 N0776118 N0776116 N0777901 N0774095 N0777691
PERCN0777156EA 204 EUR
PERCN0777156 PERCN0777159 PERCN0777167 PERCN0777168 PERCN9301205 PERCB0193233 PERCB0193234 PERCB0508901 PERCN0777701 PERCN0777702 PERCN0777703 PERCN0777704 PERCN0777705 PERCN0777706 PERCN0777692 PERCN0777693 PERCN0777695 PERCN0777696 PERCN0777697 PERCN0777698 PERCN0777699 PERCN0777700 PERCN0777940 PERCN0777694 PERCN0776118 PERCN0776116 PERCN0777901 PERCN0774095 PERCN0777691
Test tubes, for autosamplers
Buisjes Reageerbuisjes
Translucent polypropylene tubes are designed to provide excellent chemical resistance.
  • Round or conical bottom tubes
  • With or without caps
PerformR™ conical tubes are designed for routine everyday applications and storage needs.
SuperClear™ tubes are made from a unique medical grade resin.
PerformR and SuperClear tubes are available with either a flat cap with a two stage seal, or a plug style cap that includes a very deep sealing area.