Inlet liners
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03300905 N6101539 N6101703 N6121001 N6121003 N6121004 N6121006 N6101052 N6101372 N6121020 N6121022 N6502000 N6502002 N6502003 N6502004 N6502005 N6502006 N6502007 N6502009 N6502010 N6502011 N6502012 N6502017 N9302949 N1011447 N6101375 N6121002
PERC03300905EA 96.9 EUR
PERC03300905 PERCN6101539 PERCN6101703 PERCN6121001 PERCN6121003 PERCN6121004 PERCN6121006 PERCN6101052 PERCN6101372 PERCN6121020 PERCN6121022 PERCN6502000 PERCN6502002 PERCN6502003 PERCN6502004 PERCN6502005 PERCN6502006 PERCN6502007 PERCN6502009 PERCN6502010 PERCN6502011 PERCN6502012 PERCN6502017 PERCN9302949 PERCN1011447 PERCN6101375 PERCN6121002
Inlet liners
Benodigdheden voor chromatografie Injectiepoorten voor chromatografiesystemen
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
  • Liners packed with wool help to trap non-volatile material