Nickel standard solution, for ICP-MS, Pure Plus grade
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9303747
PERCN9303747EA 60.2 EUR
PERCN9303747
Nickel standard solution, for ICP-MS, Pure Plus grade
Standaarden Kationische standaarden Nikkel standaarden
Single element standard is supplied with a comprehensive Certificate of Analysis that documents the quality and reliability.
  • Analysed by ICP-OES
  • Analysed by classical wet assay
  • 67 trace impurities analysed by ICP-MS of the final solution and reported on the certificate
  • Impurities reported at ppb level
  • Prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA
Opgelet: These standards are in acid solution, for full details of hazards please read the accompanying Material Safety Data Sheets (MSDS).