Capillary columns, Elite-Volatiles for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9316388 N9316390 N9316391 N9316393
PERCN9316388EA 4390 EUR
PERCN9316388 PERCN9316390 PERCN9316391 PERCN9316393
Capillary columns, Elite-Volatiles for autosystem
Kolommen voor chromatografie
Elite Volatiles specialty phase capillary columns are ideal for the analysis of volatile organic compounds.
Order Now
Specificaties
Environmentally Preferable