Capillary Columns, Elite-WAX ETR for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9316547 N9316549 N9316550 N9316551 N9316552 N9316554 N9316555 N9316556 N9316557 N9316558 N9316560 N9316561 N9316567 N9316569 N9316570 N9316571
PERCN9316547EA 996 EUR
PERCN9316547 PERCN9316549 PERCN9316550 PERCN9316551 PERCN9316552 PERCN9316554 PERCN9316555 PERCN9316556 PERCN9316557 PERCN9316558 PERCN9316560 PERCN9316561 PERCN9316567 PERCN9316569 PERCN9316570 PERCN9316571
Capillary Columns, Elite-WAX ETR for autosystem
Kolommen voor chromatografie
Elite-WAX ETR Polyethylene Glycol capillary columns are ideal for the analysis of polar compounds.