Capillary columns, Elite-1 for autosystem
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N9316025 N9316030 N9316058 N9316005 N9316006 N9316007 N9316008 N9316010 N9316011 N9316013 N9316014 N9316015 N9316017 N9316019 N9316021 N9316022 N9316023 N9316024 N9316026 N9316028 N9316029 N9316031 N9316043 N9316046 N9316047 N9316048 N9316049 N9316050 N9316051 N9316053 N9316054 N9316063 N9316065 N9316580
PERCN9316025EA 1450 EUR
PERCN9316025 PERCN9316030 PERCN9316058 PERCN9316005 PERCN9316006 PERCN9316007 PERCN9316008 PERCN9316010 PERCN9316011 PERCN9316013 PERCN9316014 PERCN9316015 PERCN9316017 PERCN9316019 PERCN9316021 PERCN9316022 PERCN9316023 PERCN9316024 PERCN9316026 PERCN9316028 PERCN9316029 PERCN9316031 PERCN9316043 PERCN9316046 PERCN9316047 PERCN9316048 PERCN9316049 PERCN9316050 PERCN9316051 PERCN9316053 PERCN9316054 PERCN9316063 PERCN9316065 PERCN9316580
Capillary columns, Elite-1 for autosystem
Kolommen voor chromatografie
The Elite-1 columns are 100% dimethyl polysiloxane and come in a variety of sizes.