Push brooms, soft sweep
Leverancier: PERFEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2624B
PERF2624BEA 52.9 EUR
PERF2624B
Push brooms, soft sweep
Bezems
These brooms are made of extremely durable polypropylene fibres which are non-conductive and will not absorb bacteria, liquids, or odours.
This specially formulated polypropylene fill is excellent for sweeping very fine and medium particles from resilient smooth floors, vinyl, linoleum, terrazzo, marble, tile, finished wood and smooth concrete.
Bestelinformatie: Polymer adjustable or fixed length handles must be ordered separately.