Reversed phase HPLC columns, Kinetex® C18
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
00B-4462-AN 00F-4462-AN 00F-4475-AN 00F-4462-E0 00D-4462-E0 00B-4462-Y0 00B-4475-AN 00B-4462-E0 00A-4462-AN 00A-4462-E0 00C-4462-E0 00C-4462-Y0 00D-4462-AN 00D-4462-Y0 00D-4475-AN 00F-4462-Y0 00C-4462-AN 00A-4475-AN 00B-4515-AN 00B-4601-AN 00F-4601-E0 00G-4601-E0 00D-4601-AN 00F-4601-AN
PHEN00B-4462-ANEA 0 EUR
PHEN00B-4462-AN PHEN00F-4462-AN PHEN00F-4475-AN PHEN00F-4462-E0 PHEN00D-4462-E0 PHEN00B-4462-Y0 PHEN00B-4475-AN PHEN00B-4462-E0 PHEN00A-4462-AN PHEN00A-4462-E0 PHEN00C-4462-E0 PHEN00C-4462-Y0 PHEN00D-4462-AN PHEN00D-4462-Y0 PHEN00D-4475-AN PHEN00F-4462-Y0 PHEN00C-4462-AN PHEN00A-4475-AN PHEN00B-4515-AN PHEN00B-4601-AN PHEN00F-4601-E0 PHEN00G-4601-E0 PHEN00D-4601-AN PHEN00F-4601-AN
Reversed phase HPLC columns, Kinetex® C18
Kolommen voor chromatografie
Kinetex C18 are core-shell silica supported 100 Å columns with TMS endcapping for general reversed phase separations.
  • Suitable for ultra-high performance for hydrophobic compounds
  • High particle density helps create optimal bed structure which reduces band broadening effects of Eddy diffusion
  • Ultra-high performance on UHPLC systems
  • Reduced diffusion path improves efficiency