Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
00D-4256-B0 00F-4256-B0 00B-4256-B0 00G-4257-E0 00B-4256-Y0 00F-4257-Y0 00D-4256-E0 00B-4256-A0 00F-4256-E0 00F-4257-E0
PHEN00D-4256-B0EA 0 EUR
PHEN00D-4256-B0 PHEN00F-4256-B0 PHEN00B-4256-B0 PHEN00G-4257-E0 PHEN00B-4256-Y0 PHEN00F-4257-Y0 PHEN00D-4256-E0 PHEN00B-4256-A0 PHEN00F-4256-E0 PHEN00F-4257-E0
Luna® Phenyl-Hexyl Reversed Phase HPLC Columns
Kolommen voor chromatografie
Luna® Phenyl-Hexyl provides separations not achievable on C18 or C8 columns; such as increased retention for polar, aromatic compounds as well as reversals in analyte elution order. Luna® Phenyl-Hexyl is a reproducible, extremely stable phenyl phase. Most phenyl phases use a short propyl (3 carbon) linker, which limits phase stability.
  • Highly reproducible and stable phenyl phase
  • Dual selectivity of both phenyl phase and a short alkyl phase (C5 or C8)
  • Excellent retention of aromatic and polar, amine compounds
The Phenyl-Hexyl bonded phase employs aMore Product Information
Order Now
Specificaties
Environmentally Preferable
Koolstofgehalte
Endcapping
pH-bereik
Oppervlakte
USP code