Normal phase chromatography columns, Luna® CN
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
00F-4255-E0 00F-4255-Y0 00G-4254-E0 00G-4255-E0
PHEN00F-4255-E0EA 0 EUR
PHEN00F-4255-E0 PHEN00F-4255-Y0 PHEN00G-4254-E0 PHEN00G-4255-E0
Normal phase chromatography columns, Luna® CN
Kolommen voor chromatografie
Luna® columns are ideal for separation of polar compounds.
These are fully porous silica supported columns.